Learn More
Meso Scale Discovery UPLEX HUM LIGHT/TNFSF14 ASYQP
SDP
Supplier:  Meso Scale Discovery K151AHCK21
Encompass
The Meso Scale Discovery U-PLEX Human Light TNF Superfamily Member 14 (TNFSF14) Assay is a multiplex immunoassay that measures human TNFSF14 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption This assay allows for the quantitative measurement of TNFSF14 levels with high precision and accuracy It requires 50 L or less of sample per well and can perform up to 70 assays on a single 96-well plate The U-PLEX platform enables efficient and flexible multiplexing while delivering reliable results
Catalog No. 50-000-81658