Learn More
Meso Scale Discovery UPLEX HUM MCP3 ASY384well
SDP
Supplier:  Meso Scale Discovery K251XIK4
Encompass
The Meso Scale Discovery U-PLEX Human MCP-3 (384-well) Assay Kit is a 384-well multiplex immunoassay that measures human Monocyte Chemoattractant Protein 3 (MCP-3) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires a Meso Scale Discovery SECTOR Imager instrument to read the plate The kit includes a 384-well carbon electrode plate coated with capture antibodies detection antibodies calibrators and controls needed to perform the assay
Catalog No. 50-000-80668
Only null left
Add to Cart
Request a Quote