Learn More
Meso Scale Discovery UPLEX HUM MCP3 ASYQP 25 PL
SDP
Supplier:  Meso Scale Discovery K151XIK24
Encompass
The U-PLEX Human MCP-3 Assay (K151XIK) is a multiplex immunoassay platform from Meso Scale Discovery for quantitative measurement of MCP-3 (Monocyte Chemoattractant Protein-3) in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The U-PLEX technology enables researchers to measure cytokines and biomarkers with superior precision for applications in immunology inflammation research and drug development studies
Catalog No. 50-000-79632
Only null left
Add to Cart
Request a Quote