Learn More
Meso Scale Discovery UPLEX HUM MIF ASY384well
SDP
Supplier:  Meso Scale Discovery K251XJK4
Encompass
The Meso Scale Discovery U-PLEX Human MIF 384-well Assay is a multiplex immunoassay kit for the quantitative measurement of macrophage migration inhibitory factor (MIF) in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and is designed for use on the MESO QuickPlex SQ 120 instrument The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG reagents calibrators and an assay diluent The assay has a typical sensitivity of 0 08 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-80648
Only null left
Add to Cart
Request a Quote