Learn More
Meso Scale Discovery UPLEX HUM MIP1B ASY384well
SDP
Supplier:  Meso Scale Discovery K251UKK2
Encompass
The Meso Scale Discovery U-PLEX Human MIP-1 384-Well Assay Kit is a multiplex immunoassay that measures human MIP-1 in serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a 384-well plate format This assay requires a Meso Scale Discovery SECTOR Imager and provides reagents for one 384-well plate to perform up to 380 tests in duplicate
Catalog No. 50-000-79412
Only null left
Add to Cart
Request a Quote