Learn More
Meso Scale Discovery UPLEX HUM MIP3 ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K151UZK4
Encompass
The Meso Scale Discovery U-PLEX Human MIP-3alpha Assay Kit is a multiplex immunoassay that measures human MIP-3alpha in serum plasma cell culture supernatants and other biological samples It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less of sample and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to quantify MIP-3alpha concentration This assay allows rapid reproducible quantification of the chemokine MIP-3alpha from small sample volumes
Catalog No. 50-000-79034
Add to Cart
Request a Quote