Learn More
Meso Scale Discovery UPLEX HUM MMP9 total ASYQP
SDP
Supplier:  Meso Scale Discovery K151AHMK21
Encompass
The Meso Scale Discovery U-PLEX Human MMP-9 Total Assay is an electrochemiluminescent immunoassay for the quantitative measurement of total MMP-9 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results in just 3 5 hours The assay has a lower limit of detection of 0 03 ng/mL and an upper limit of quantification of 10 000 ng/mL It includes a 96-well plate pre-coated with capture antibody detection antibody calibrator and sample diluent This assay allows high-throughput and accurate quantification of MMP-9 levels
Catalog No. 50-000-79556