Learn More
Meso Scale Discovery UPLEX HUM Nectin4 ASYQP
SDP
Supplier:  Meso Scale Discovery K151AHFK24
Encompass
The Meso Scale Discovery U-PLEX Human Nectin-4 Assay is a multiplex immunoassay kit for measuring Nectin-4 in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls buffers and seals It measures Nectin-4 concentrations from 0 05-500 ng/mL in about 3 5 hours aiding research on Nectin-4 as a cancer biomarker
Catalog No. 50-000-78399
Add to Cart
Request a Quote