Learn More
Meso Scale Discovery UPLEX HUM NGAL/LCN2 ASY
SDP
Supplier:  Meso Scale Discovery K151Z1K4
Encompass
The Meso Scale Discovery U-PLEX Human NGAL (Lipocalin-2) Assay is a multiplex immunoassay kit that measures natural and recombinant human Neutrophil Gelatinase-Associated Lipocalin (NGAL) in serum plasma urine cell culture supernatants and other biological samples It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid results This assay allows for the quantitative measurement of NGAL which is a biomarker of kidney injury It requires low sample volume and has high reproducibility The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator and controls
Catalog No. 50-000-80099
Only null left
Add to Cart
Request a Quote