Learn More
Meso Scale Discovery UPLEX HUM OX40/TNFRSF4 ASY384
SDP
Supplier:  Meso Scale Discovery K251T7K4
Encompass
The Meso Scale Discovery U-PLEX Human OX40 (TNFRSF4) 384-well Assay is a multiplex immunoassay kit for quantifying OX40 and other biomarkers in human serum plasma and cell culture supernatants It uses electrochemiluminescent detection in a 384-well format offering high sensitivity and a broad dynamic range The assay requires 25 L of sample and takes less than 3 5 hours Each kit includes reagents for one 384-well plate such as antibody-coated plates calibrator diluents detection antibodies read buffer and wash buffer allowing customizable panels for measuring OX40 and other biomarkers
Catalog No. 50-000-80726
Add to Cart
Request a Quote