Learn More
Meso Scale Discovery UPLEX HUM OX40/TNFRSF4 ASYQP
SDP
Supplier:  Meso Scale Discovery K151T7K24
Encompass
The Meso Scale Discovery U-PLEX Human OX40 (TNFRSF4) Assay Kit is a multiplex immunoassay for measuring OX40 in human serum plasma and cell culture supernatant It uses electrochemiluminescent detection offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG buffers diluent calibrators and controls It is for research use only not diagnostics enabling efficient and accurate OX40 quantification
Catalog No. 50-000-78564
Add to Cart
Request a Quote