Learn More
Meso Scale Discovery UPLEX HUM OX40/TNFRSF4 ASYQP
SDP
Supplier:  Meso Scale Discovery K151T7K22
Encompass
The Meso Scale Discovery U-PLEX Human OX40 (TNFRSF4) Assay Kit is a multiplex immunoassay that measures OX40 in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay kit includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and controls The assay has a typical sensitivity of 0 23 pg/mL and an upper limit of quantification of 10 000 pg/mL It requires 50 L of sample and can perform up to 80 tests in the provided 96-well plate
Catalog No. 50-000-81357