Learn More
Meso Scale Discovery UPLEX HUM PD1 epitope 1 ASY
SDP
Supplier:  Meso Scale Discovery K151U7K4
Encompass
The Meso Scale Discovery U-PLEX Human PD1 Epitope 1 Assay Kit is a multiplex immunoassay for measuring PD1 epitope 1 in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection for high sensitivity broad dynamic range and precision The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators and controls It quantifies PD1 epitope 1 from 0 01 ng/mL to 1000 ng/mL enabling accurate measurement for research
Catalog No. 50-000-78999
Only null left
Add to Cart
Request a Quote