Learn More
Meso Scale Discovery UPLEX HUM PD1 epitope 2 ASYQP
SDP
Supplier:  Meso Scale Discovery K151V7K22
Encompass
The Meso Scale Discovery U-PLEX Human PD1 Epitope 2 Assay is a multiplex immunoassay kit for detecting and quantifying PD1 epitope 2 in human serum plasma and cell culture supernatant It uses MSD s electrochemiluminescent technology offering high sensitivity a dynamic range of up to 5 logs and compatibility with small sample volumes The assay monitors immune checkpoint inhibitor therapy response and includes a plate with capture and detection antibodies calibrators and controls An MSD instrument like QuickPlex is required to read the plate
Catalog No. 50-000-78997