Learn More
Meso Scale Discovery UPLEX HUM PDepitope 2 ASY384
SDP
Supplier:  Meso Scale Discovery K251V7K2
Encompass
The U-PLEX Human PD1 Epitope 2 384-Well Assay Kit from Meso Scale Discovery is a 384-well immunoassay that measures PD1 epitope 2 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This assay kit includes a plate coated with anti-PD1 capture antibody detection antibody read buffer wash buffer blocker reagents calibrator and controls needed to perform the immunoassay It allows for the quantitative measurement of PD1 epitope 2 levels with a testing range of 50-50 000 pg/mL
Catalog No. 50-000-79853
Add to Cart
Request a Quote