Learn More
Meso Scale Discovery UPLEX HUM PDL1 epitope 1 ASYQP
SDP
Supplier:  Meso Scale Discovery K151Z7K22
Encompass
The Meso Scale Discovery U-PLEX Human PD-L1 (Epitope 1) Assay Kit is a multiplex immunoassay that measures PD-L1 levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay kit includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and diluent It allows quantification of PD-L1 levels which can help monitor immune checkpoint inhibitor therapy response and progression in cancer patients The kit has high lot-to-lot consistency and requires low sample volume
Catalog No. 50-000-79216