Learn More
Meso Scale Discovery UPLEX HUM PDL1 epitope 2 ASY
SDP
Supplier:  Meso Scale Discovery K151AHSK2
Encompass
The Meso Scale Discovery U-PLEX Human PD-L1 Epitope 2 Assay is a multiplex immunoassay kit for detecting and quantifying PD-L1 and other biomarkers in human samples It uses MSD s electrochemiluminescent technology offering high sensitivity a dynamic range of up to 5 logs and compatibility with serum plasma and cell culture supernatants The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls read buffer sample diluent and plate seals facilitating efficient measurement of biomarkers for cancer immunotherapy research
Catalog No. 50-000-78860