Learn More
Meso Scale Discovery UPLEX HUM PDL1 epitope 2 ASYQP
SDP
Supplier:  Meso Scale Discovery K151AHSK21
Encompass
The Meso Scale Discovery U-PLEX Human PD-L1 Epitope 2 Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of PD-L1 and other biomarkers from human samples It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and compatibility with a variety of sample types including serum plasma and cell culture supernatants This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators to generate a standard curve It allows for efficient and reproducible analysis of PD-L1 expression alongside other immune checkpoint biomarkers
Catalog No. 50-000-78861