Learn More
Meso Scale Discovery UPLEX HUM proMMP9 ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K151AHLK4
Encompass
The Meso Scale Discovery U-PLEX Human ProMMP-9 Assay is a multiplex immunoassay for measuring ProMMP-9 in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The assay detects ProMMP-9 as low as 45 pg/mL with just 25 L of sample It includes a plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrators for a standard curve The assay takes about 3 5 hours providing quantitative ProMMP-9 levels for research
Catalog No. 50-000-80475
Add to Cart
Request a Quote