Learn More
Meso Scale Discovery UPLEX HUM PYY TTL ASY384well
SDP
Supplier:  Meso Scale Discovery K2516BK2
Encompass
The Meso Scale Discovery U-PLEX Human PYY Total 384-Well Assay is an electrochemiluminescent immunoassay for the quantitative measurement of total Peptide YY (PYY) in human serum plasma and cell culture supernatants This 384-well multiplex assay kit utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours with a lower limit of detection of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL The assay measures both PYY(1-36) and PYY(3-36) and includes reagents for 384 tests in a ready-to-use room temperature stable format
Catalog No. 50-000-78462
Only null left
Add to Cart
Request a Quote