Learn More
Meso Scale Discovery UPLEX HUM RANKL/TNFSF11 ASY
SDP
Supplier:  Meso Scale Discovery K151M7K4
Encompass
The Meso Scale Discovery U-PLEX Human RANKL (TNFSF11) Assay Kit is a multiplex immunoassay for measuring RANKL in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity broad dynamic range and precision The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators and controls Sensitivity is 0 04 pg/mL with an upper quantification limit of 10 000 pg/mL enabling accurate RANKL quantitation for research like biomarker discovery and validation
Catalog No. 50-000-81179
Add to Cart
Request a Quote