Learn More
Meso Scale Discovery UPLEX HUM RANKL/TNFSF11 ASYQP
SDP
Supplier:  Meso Scale Discovery K151M7K22
Encompass
The Meso Scale Discovery U-PLEX Human RANKL (TNFSF11) Assay Kit is an electrochemiluminescent immunoassay for measuring RANKL in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive results in 3 5 hours The kit includes a 96-well plate with an anti-RANKL capture antibody a detection antibody with an MSD SULFO-TAG calibrators and a diluent The assay range is 2 4-10 000 pg/mL with no cross-reactivity enabling accurate RANKL quantitation for bone metabolism research
Catalog No. 50-000-80799