Learn More
Meso Scale Discovery UPLEX HUM S100A12 ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K151AHPK4
Encompass
The Meso Scale Discovery U-PLEX Human S100A12 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of S100A12 and other biomarkers in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology for high sensitivity and broad dynamic range The assay requires 50 L or less sample volume and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator The assay has a typical lower limit of detection of 0 04 ng/mL and an upper limit of 10 000 ng/mL for S100A12 It allows high-throughput analysis of S100A12 levels
Catalog No. 50-000-79091
Add to Cart
Request a Quote