Learn More
Meso Scale Discovery UPLEX HUM Serpin A1 ASY384well
SDP
Supplier:  Meso Scale Discovery K251W9K2
Encompass
The Meso Scale Discovery U-PLEX Human Serpin A1 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of Serpin A1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides a 384-well plate formatted assay for high throughput analysis of up to 80 samples The assay has a typical sensitivity of 0 08 ng/mL and requires just 50 L of sample It includes a plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator and controls This assay kit offers a rapid sensitive and reproducible method for measuring Serpin A1 levels in biological samples
Catalog No. 50-000-79474
Only null left
Add to Cart
Request a Quote