Learn More
Meso Scale Discovery UPLEX HUM sTfR1 ASY384well
SDP
Supplier:  Meso Scale Discovery K251P9K4
Encompass
The U-PLEX Human STFR 1-384 Well Assay from Meso Scale Diagnostics is a multiplex immunoassay for measuring up to 10 human proteins per well It uses electrochemiluminescence detection in a 384-well format offering high sensitivity and minimal sample use Requiring 10-25 L per well it is compatible with MSD platforms and allows custom analyte panels from over 175 biomarkers The kit includes a 384-well plate detection antibodies read buffer sample diluent calibrators and controls
Catalog No. 50-000-80698
Add to Cart
Request a Quote