Learn More
Meso Scale Discovery UPLEX HUM Synuclein
SDP
Supplier:  Meso Scale Discovery K151WKK4
Encompass
The Meso Scale Discovery U-PLEX Human Alpha-Synuclein Kit is an electrochemiluminescent immunoassay for the quantitative measurement of alpha-synuclein in human serum plasma and CSF The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and assay diluent The assay has a typical sensitivity of 0 064 pg/mL and an upper limit of quantification of 2500 pg/mL The total assay time is approximately 3 5 hours This highly sensitive and reproducible assay allows for the accurate quantification of alpha-synuclein levels which may aid in understanding synucleinopathies such as Parkinson s disease
Catalog No. 50-000-81834
Add to Cart
Request a Quote