Learn More
Meso Scale Discovery UPLEX HUM TGFB2 ASY384well
SDP
Supplier:  Meso Scale Discovery K251XUK2
Encompass
The Meso Scale Discovery U-PLEX Human TGF-beta2 384-Well Assay Kit is a multiplex immunoassay that measures TGF-beta2 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This 384-well plate assay allows for efficient and high-throughput analysis of up to 80 samples in duplicate It includes a plate coated with capture antibodies detection antibodies Read Buffer and calibrator The kit has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for TGF-beta2
Catalog No. 50-000-80669
Add to Cart
Request a Quote