Learn More
Meso Scale Discovery UPLEX HUM TIGIT ASY384well
SDP
Supplier:  Meso Scale Discovery K251W7K4
Encompass
The Meso Scale Discovery U-PLEX Human TIGIT 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of the TIGIT protein in human serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a 384-well plate format allowing high-throughput analysis of up to 80 samples The kit includes a TIGIT-specific capture antibody a ruthenium-labeled detection antibody and calibrators to generate a standard curve The assay has a typical sensitivity of 0 3 ng/mL and an upper limit of quantification of 2500 pg/mL for TIGIT The kit is for research use only
Catalog No. 50-000-80516
Only null left
Add to Cart
Request a Quote