Learn More
Meso Scale Discovery UPLEX HUM TLR1 ASY384well
SDP
Supplier:  Meso Scale Discovery K251J7K4
Encompass
The U-PLEX Human TLR1 384-Well Assay from Meso Scale Discovery is a multiplex immunoassay that measures human Toll-like receptor 1 (TLR1) in cell and tissue lysates It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and minimal sample consumption in a 384-well plate format This assay requires only 2-10 L of sample per well and can measure TLR1 across a range of 0 11 - 600 ng/mL It includes reagents for running 384 sample wells along with assay diluent calibrator and quality controls This quantitative immunoassay allows high-throughput profiling of TLR1 which plays a key role in innate immunity
Catalog No. 50-000-80732
Add to Cart
Request a Quote