Learn More
Meso Scale Discovery UPLEX HUM TNF ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K151UCK4
Encompass
The Meso Scale Discovery U-PLEX Human TNF-alpha Assay Kit is a multiplex immunoassay for measuring human TNF-alpha in various biological samples It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The assay offers high specificity and low cross-reactivity It includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags calibrators and buffer It measures TNF-alpha and other biomarkers with sensitivity of 0 04 pg/mL and an upper limit of 10 000 pg/mL
Catalog No. 50-000-79119
Add to Cart
Request a Quote