Learn More
Meso Scale Discovery UPLEX HUM TNF ASY384well
SDP
Supplier:  Meso Scale Discovery K251UCK4
Encompass
The Meso Scale Discovery U-PLEX Human TNF-alpha 384-Well Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of TNF-alpha in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide a highly sensitive and reproducible assay in a 384-well format The kit includes a TNF-alpha calibrator and all necessary reagents and plates for performing the assay with a 384-well plate reader The assay has a typical sensitivity of 0 05 pg/mL and a broad dynamic range from 0 05-1000 pg/mL It requires a low sample volume of 25 L and provides results in as little as 3 5 hours
Catalog No. 50-000-80465
Only null left
Add to Cart
Request a Quote