Learn More
Meso Scale Discovery UPLEX HUM TNFRII ASYQP 25 PL
SDP
Supplier:  Meso Scale Discovery K151AHRK24
Encompass
The Meso Scale Discovery U-PLEX Human TNF RII Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human TNF RII in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and controls It allows sensitive reproducible and rapid quantification of human TNF RII levels for life science research applications
Catalog No. 50-000-80601
Add to Cart
Request a Quote