Learn More
Meso Scale Discovery UPLEX HUM VCAM1 ASYQP 25 PL
SDP
Supplier:  Meso Scale Discovery K151H9K24
Encompass
The Meso Scale Discovery U-PLEX Human VCAM-1 Assay Kit quantifies VCAM-1 in human serum plasma and cell culture supernatants using electrochemiluminescent detection It employs MSD GOLD Streptavidin Multi-Array 96-well plates and a VCAM-1 capture antibody A SULFO-TAG labeled detection antibody identifies the analyte Electricity applied to electrodes causes SULFO-TAG to emit light measured by an MSD instrument The kit includes a VCAM-1 calibrator and control for standard curve and assay validation detecting VCAM-1 from 0 05-1000 ng/mL
Catalog No. 50-000-81151
Only null left
Add to Cart
Request a Quote