Learn More
Meso Scale Discovery UPLEX HUM VEGFR1/FltASY384well
SDP
Supplier:  Meso Scale Discovery K251AHHK2
Encompass
The Meso Scale Discovery U-PLEX Human VEGFR-1 (FLT-1) Assay is a 384-well multiplex immunoassay kit for quantifying human VEGFR-1 in cell culture supernatants serum plasma and other samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a VEGF-A capture antibody a VEGFR-1 detection antibody and calibrators and controls It measures VEGFR-1 concentrations from 50 to 100 000 pg/mL suitable for studying angiogenesis and related biology
Catalog No. 50-000-80748
Only null left
Add to Cart
Request a Quote