Learn More
Meso Scale Discovery UPLEX HUM YKL40 ASY384well
SDP
Supplier:  Meso Scale Discovery K251VLK2
Encompass
The Meso Scale Discovery U-PLEX Human YKL-40 384-Well Assay is an electrochemiluminescent immunoassay designed for the quantitative measurement of YKL-40 in human serum plasma (EDTA heparin) and cell culture supernatants This 384-well plate assay utilizes MSD s proprietary SULFO-TAG labels and provides a sensitive reproducible and high-throughput method for measuring YKL-40 with a lower limit of detection of 0 31 ng/mL and an upper limit of 10 000 ng/mL The assay requires a low sample volume of 25 L and the total assay time is less than 3 5 hours This product can aid in understanding YKL-40 s role in diseases involving chronic inflammation and tissue remodeling
Catalog No. 50-000-80976
Only null left
Add to Cart
Request a Quote