Learn More
Meso Scale Discovery U-PLEX Human GLP-1 (inactive) Assay, SECTOR (25 PL)
SDP
Supplier:  Meso Scale Discovery K1515VK4
Encompass
"The U-PLEX platform offers validated components that can be combined to create high quality assays quickly and easily. U-PLEX Assay has the essential components for an assay that measures single-analyte format. For information on multiplexing with other analytes, please see U-PLEX Custom Metabolic Group 1 (human) Assays. Related Key Terms:Glucagon-Like Peptide-1, GLP-1, metabolic diseases, metabolism, proglucagon, insulin, diabetes"
Catalog No. NC3786658
Only null left
Add to Cart
Request a Quote