Learn More
Meso Scale Discovery U-PLEX Human Glucagon Assay, QuickPlex (5 PL)
SDP
Supplier:  Meso Scale Discovery K1515YK22
Encompass
"The U-PLEX platform offers validated components that can be combined to create high quality assays quickly and easily. U-PLEX Human Glucagon Assay has the essential components for an assay that measures Glucagon in a single-analyte format. For information on multiplexing Glucagon with other analytes, please see U-PLEX Custom Metabolic Group 1 (human) Assays. Related Key Terms:GCG, metabolism, metabolic, gluconeogenesis, glycogenolysis, ketogenesis, glucose, Metabolic Syndrome, diabetes"
Catalog No. NC3786859