Learn More
Meso Scale Discovery U-PLEX Human TSLP Antibody Set B21WH-3, 5 Plate Size
SDP
Supplier:  Meso Scale Discovery B21WH3
Encompass
The U-PLEX platform offers validated components that can be combined to create high quality assays quickly and easily. The U-PLEX Human TSLP Antibody Set is a matched pair of antibodies that can be used (with other U-PLEX components such as plates, calibrators, diluents, and MSD GOLD Read Buffer B) to build U-PLEX assays that measure TSLP. These assays can measure TSLP alone or in multiplex panels with other biomarkers.
Catalog No. NC3502327