Learn More
Meso Scale Discovery UPLEX Interferon Combo ms
SDP
Supplier:  Meso Scale Discovery K15320K4
Encompass
The U-PLEX Interferon Combo 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay for measuring mouse interferon alpha beta and gamma in various samples It includes a 96-well plate with capture and detection antibodies Read Buffer and calibrator The assay using MSD s MULTI-ARRAY technology takes about 3 5 hours and detects 0 05-2500 pg/mL It offers high sensitivity excellent data quality and low sample volume requirements optimized for mouse samples
Catalog No. 50-000-81881
Only null left
Add to Cart
Request a Quote