Learn More
Meso Scale Discovery UPLEX Macrophage M2 Combo hu
SDP
Supplier:  Meso Scale Discovery K15337K2
Encompass
The U-PLEX Macrophage M2 Combo 1 Human Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of human IL-10 CCL17 CCL18 and CCL22 in cell culture supernatants serum plasma and other biological samples The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer and calibrator The electrochemiluminescent assay takes approximately 3 5 hours to complete and has a detection range of 0 05-2500 pg/mL This kit enables quantification of M2 macrophage activation
Catalog No. 50-000-81884
Only null left
Add to Cart
Request a Quote