Learn More
Meso Scale Discovery UPLEX Macrophage MCombo hu
SDP
Supplier:  Meso Scale Discovery K15336K4
Encompass
The U-PLEX Macrophage M1 Combo 1 Human Kit from Meso Scale Discovery is a multiplex immunoassay kit that enables simultaneous quantification of 10 biomarkers related to M1 macrophage activation in human samples The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer It utilizes electrochemiluminescence for high sensitivity and broad dynamic range This kit allows efficient profiling of pro-inflammatory M1 macrophages
Catalog No. 50-000-81515
Only null left
Add to Cart
Request a Quote