Learn More
Meso Scale Discovery UPLEX Macrophage MCombo ms
SDP
Supplier:  Meso Scale Discovery K15408K2
Encompass
The U-PLEX Macrophage M1 Combo 1 by Meso Scale Diagnostics is a multiplex assay kit for measuring human IFN-gamma IL-1beta IL-6 IL-8 IL-10 IL-12p70 and TNF-alpha It uses electrochemiluminescent detection on Meso Scale instruments Monoclonal antibodies form a sandwich complex on carbon electrodes and a label solution produces light proportional to analyte levels This assay efficiently analyzes multiple inflammatory cytokines related to M1 macrophage activation from various biological samples
Catalog No. 50-000-79764
Only null left
Add to Cart
Request a Quote