Learn More
Meso Scale Discovery UPLEX Metabolic 2Plex 2 rat
SDP
Supplier:  Meso Scale Discovery K15313K2
Encompass
The U-PLEX Metabolic 2-Plex Combo 2 Rat is a multiplex immunoassay kit from Meso Scale Discovery that allows simultaneous quantitative measurement of insulin and leptin in rat serum plasma and cell culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive reproducible results with low sample volume requirements This 2-plex panel enables efficient analysis of metabolic biomarkers related to energy regulation and body weight homeostasis The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents sufficient for one 96-well plate
Catalog No. 50-000-82129