Learn More
Meso Scale Discovery UPLEX Metabolic 2Plex 2
SDP
Supplier:  Meso Scale Discovery K15303K1
Encompass
The U-PLEX Metabolic 2-plex Combo 2 Mouse is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous quantitative measurement of insulin and leptin in mouse serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology for high sensitivity and broad dynamic range The assay requires 50 L sample volume and can be read on MESO QuickPlex SQ 120 MESO SECTOR S 600 or MESO SECTOR S 6000 instruments The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents needed to complete the assay Results are obtained in as little as 3 5 hours
Catalog No. 50-000-80415