Learn More
Meso Scale Discovery UPLEX Metabolic 2Plex rat
SDP
Supplier:  Meso Scale Discovery K15312K1
Encompass
The U-PLEX Metabolic 2-plex Combo 1 Rat is a multiplex immunoassay kit from Meso Scale Diagnostics for measuring insulin and leptin in rat serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results with low sample volume This 2-plex panel aids in analyzing metabolic biomarkers related to energy regulation and obesity in preclinical rat models The kit includes a 96-well plate capture and detection antibodies calibrators controls buffers and seals for one plate
Catalog No. 50-000-82440