Learn More
Meso Scale Discovery UPLEX Metabolic 3Plex 2 rat
SDP
Supplier:  Meso Scale Discovery K15315K4
Encompass
The U-PLEX Metabolic 3-Plex Combo 2 Rat kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring insulin leptin and resistin in rat samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volumes This panel aids in analyzing metabolic biomarkers related to obesity diabetes and insulin resistance in preclinical rat studies The kit includes a 96-well plate with necessary reagents for one plate
Catalog No. 50-000-79141
Add to Cart
Request a Quote