Learn More
Meso Scale Discovery UPLEX Metabolic Combo 1 rt
SDP
Supplier:  Meso Scale Discovery K15308K4
Encompass
The U-PLEX Metabolic Combo 1 Rat Kit from Meso Scale Discovery is a multiplex immunoassay kit for the simultaneous quantitative measurement of insulin leptin and resistin in rat serum plasma and tissue culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators and buffers needed to run the assay The electrochemiluminescent detection allows for high sensitivity and a broad dynamic range This kit allows for the efficient analysis of metabolic biomarkers related to obesity diabetes and metabolic syndrome in rat samples
Catalog No. 50-000-81873
Add to Cart
Request a Quote