Learn More
Meso Scale Discovery UPLEX Metabolic Hormone
SDP
Supplier:  Meso Scale Discovery K15316K4
Encompass
The Meso Scale Discovery U-PLEX Metabolic Hormones Combo 1 Rat Kit is a multiplex immunoassay for measuring insulin leptin and resistin in rat serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible quantification from small samples The kit includes a 96-well plate with capture and detection antibodies calibrators controls buffers and seals for one plate aiding research in obesity diabetes and metabolic syndrome
Catalog No. 50-000-82492
Only null left
Add to Cart
Request a Quote