Learn More
Meso Scale Discovery UPLEX Metastasis Combo 1HMN
SDP
Supplier:  Meso Scale Discovery K15602K4
Encompass
The U-PLEX Metastasis Combo 1 Human Sector Kit from Meso Scale Discovery is a multiplex immunoassay kit for quantifying 10 protein biomarkers of metastasis and tumor progression in human serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies for MMP-9 TIMP-1 PAI-1 uPA VEGF-C VEGF-D Endoglin IL-6 IL-8 and MIF along with detection antibodies and calibrators Compatible with MSD instruments it allows sensitive reproducible and rapid quantification from a small sample
Catalog No. 50-000-80308
Add to Cart
Request a Quote