Learn More
Meso Scale Discovery UPLEX Mouse BAFF ASYQP 25 PL
SDP
Supplier:  Meso Scale Discovery K1523QK24
Encompass
The Meso Scale Discovery U-PLEX Mouse BAFF Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of BAFF in mouse serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and has a testing time of 3 5 hours It contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and diluent This assay kit can be used to measure BAFF levels in mouse models of autoimmune diseases
Catalog No. 50-000-81348
Add to Cart
Request a Quote